Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145146.2 | NP_660128.2 | 8 | 974 | ||
GenBank mRNA | AJ011080.1 | - | 8 | 1002 | ||
AK143987.1 | - | 8 | 975 | |||
BC026681.1 | - | 8 | 892 | |||
BC100597.1 | - | 8 | 974 | 6767654 |