Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284411.1 | NP_001271340.1 | 2 | 189 | ||
NM_001284412.1 | NP_001271341.1 | 2 | 189 | |||
NM_001284414.1 | NP_001271343.1 | 2 | 189 | |||
NM_026776.4 | NP_081052.2 | 2 | 189 | |||
GenBank mRNA | AK002500.1 | - | 2 | 151 | ||
AK003715.2 | - | 2 | 161 | |||
AK003877.1 | - | 2 | 147 | |||
AK005149.1 | - | 2 | 147 | |||
AK082933.1 | - | 2 | 168 | |||
AK145811.1 | - | 2 | 160 | |||
AK150822.1 | - | 2 | 171 | |||
AK162230.1 | - | 2 | 162 | |||
AK166008.1 | - | 2 | 168 | |||
BC029181.1 | - | 2 | 153 | 4240500 |