Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145705.3 | NP_663751.2 | 9 | 1437 | ||
GenBank mRNA | AF214013.1 | - | 9 | 1425 | ||
AF518764.1 | - | 9 | 1402 | |||
AI844697.1 | - | 1 | 299 | |||
AK011154.1 | - | 9 | 1405 | |||
AK016456.1 | - | 9 | 1371 | |||
AK028952.1 | - | 8 | 1562 | |||
AK090324.1 | - | 9 | 1404 | |||
AK169480.1 | - | 3 | 1555 | |||
AK205383.1 | - | 2 | 101 | |||
AK205420.1 | - | 2 | 98 | |||
AK214714.1 | - | 3 | 144 | |||
BC017142.1 | - | 4 | 1853 | |||
BC030347.1 | - | 9 | 1363 | 5003804 |