Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145959.1 | NP_001139431.1 | 7, 8 | 687 | ||
NM_013864.2 | NP_038892.1 | 8, 9 | 729 | |||
GenBank mRNA | AB033921.1 | - | 8, 9 | 592 | ||
AK076514.1 | - | 5, 6 | 363 | |||
AK154597.1 | - | 8, 9 | 597 | |||
AK158900.1 | - | 7, 8 | 596 | |||
AK168011.1 | - | 7, 8 | 554 | |||
AK173136.1 | - | 5, 6 | 370 | |||
BC012963.1 | - | 8, 9 | 596 | 4211421 |