Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013885.2 | NP_038913.1 | 5, 6 | 766 | ||
GenBank mRNA | AF102578.1 | - | 5, 6 | 764 | ||
AK031211.1 | - | 5, 6 | 773 | |||
AK049191.1 | - | 5, 6 | 771 | |||
AK049255.1 | - | 5, 6 | 772 | |||
AK150993.1 | - | 5, 6 | 770 | |||
AK151504.1 | - | 1, 2 | 42 | |||
AK152209.1 | - | 1, 2 | 42 | |||
AK152273.1 | - | 5, 6 | 772 | |||
AK153432.1 | - | 5, 6 | 773 | |||
AK155826.1 | - | 5, 6 | 770 | |||
AK160856.1 | - | 5, 6 | 772 | |||
AK161034.1 | - | 5, 6 | 771 | |||
AK166244.1 | - | 5, 6 | 774 | |||
AK166794.1 | - | 5, 6 | 770 | |||
AK169376.1 | - | 5, 6 | 770 | |||
AK169834.1 | - | 5, 6 | 770 | |||
AK170942.1 | - | 5, 6 | 821 | |||
AK171243.1 | - | 5, 6 | 773 | |||
AK172026.1 | - | 5, 6 | 772 | |||
AK172516.1 | - | 5, 6 | 746 | |||
AK172660.1 | - | 5, 6 | 770 | |||
BC046384.1 | - | 5, 6 | 750 | 3155573 | ||
BC052890.1 | - | 5, 6 | 754 | 30056606 | ||
CT010279.1 | - | 5, 6 | 595 |