Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083334.1 | NP_001076803.1 | 11 | 1197 | ||
NM_009668.2 | NP_033798.1 | 12 | 1290 | |||
GenBank mRNA | AK184020.1 | - | 2 | 126 | ||
AK191982.1 | - | 2 | 126 | |||
BC065160.1 | - | 11 | 1169 | 5718309 | ||
CF732768.1 | - | 3 | 327 |