Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 30, 31 | 3871 | ||
NM_001163448.1 | NP_001156920.1 | 29, 30 | 3829 | |||
NM_001163449.1 | NP_001156921.1 | 31, 32 | 3805 | |||
NM_001163450.1 | NP_001156922.1 | 30, 31 | 3781 | |||
NM_001163451.1 | NP_001156923.1 | 30, 31 | 3778 | |||
NM_001163453.1 | NP_001156925.1 | 29, 30 | 3763 | |||
NM_013931.4 | NP_038959.2 | 30, 31 | 3874 | |||
GenBank mRNA | AB005662.1 | - | 30, 31 | 3761 | ||
AB043123.1 | - | 31, 32 | 3682 | |||
AB043124.1 | - | 30, 31 | 3751 | |||
AB043125.1 | - | 30, 31 | 3748 | |||
AB093282.2 | - | 30, 31 | 3733 | |||
AF178636.1 | - | 29, 30 | 3746 | |||
AF178637.1 | - | 30, 31 | 3867 | |||
AF262046.1 | - | 30, 31 | 3857 | |||
AK039130.1 | - | 12, 13 | 1501 | |||
AK134797.1 | - | 9, 10 | 949 | |||
BC004003.1 | - | 1, 2 | 34 | |||
BC060603.1 | - | 31, 32 | 3740 | 5706805 | ||
BC065105.1 | - | 29, 30 | 3753 | 30356817 | ||
BC068312.1 | - | 30, 31 | 3852 | 30100914 |