Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027225.1 | NP_081501.1 | 11 | 2082 | ||
NM_177025.5 | NP_795999.2 | 12 | 2193 | |||
GenBank mRNA | AK037010.1 | - | 13 | 2187 | ||
AK052533.1 | - | 11 | 2031 | |||
AK135859.1 | - | 3 | 421 | |||
AK144943.1 | - | 13 | 2240 | |||
AK173076.1 | - | 11 | 1913 | |||
AY308746.1 | - | 12 | 2193 | |||
BC067007.1 | - | 11 | 2079 | 5702930 |