Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177045.3 | NP_796019.1 | 17 | 2028 | ||
GenBank mRNA | AK043839.1 | - | 17 | 2029 | ||
AK146601.1 | - | 17 | 2047 | |||
AK173271.1 | - | 13 | 1456 | |||
AK217985.1 | - | 4 | 368 | |||
BC055068.1 | - | 15 | 1670 | |||
BC062883.1 | - | 8 | 1651 | |||
BC125517.1 | - | 16 | 1918 | 40130032 | ||
BC137743.1 | - | 16 | 1918 | 8860762 |