Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252094.1 | NP_001239023.1 | 5 | 953 | ||
NM_001252095.1 | NP_001239024.1 | 5 | 934 | |||
NM_001252096.1 | NP_001239025.1 | 4 | 843 | |||
NM_001252097.1 | NP_001239026.1 | 3 | 415 | |||
NM_177101.5 | NP_796075.2 | 4 | 824 | |||
GenBank mRNA | AK044252.1 | - | 2131 | |||
AK083766.1 | - | 1793 | ||||
AK132096.1 | - | 5 | 821 | |||
BC018279.1 | - | 2 | 122 | |||
BC046911.1 | - | 3 | 354 | 6491688 | ||
BC048711.1 | - | 4 | 716 | 6328428 | ||
BC065807.1 | - | 4 | 740 | 3373106 |