Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177233.5 | NP_796207.2 | 3, 4 | 539 | ||
GenBank mRNA | AK081531.1 | - | 3, 4 | 522 | ||
AK149355.1 | - | 3, 4 | 542 | |||
AY325123.1 | - | 3, 4 | 322 | |||
BB642822.1 | - | 3, 4 | 539 | |||
BC089490.1 | - | 3, 4 | 499 | 30504398 |