Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015507.2 | NP_001015507.1 | 13, 14 | 2094 | ||
GenBank mRNA | AK122372.1 | - | 13, 14 | 2057 | ||
AK135721.1 | - | 1, 2 | 136 | |||
BC065119.1 | - | 13, 14 | 2094 | 6853519 |