Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033409.3 | NP_001028581.1 | 18 | 2018 | ||
GenBank mRNA | AK052873.1 | - | 13 | 1218 | ||
AK085901.1 | - | 18 | 1978 | |||
AK137380.1 | - | 18 | 2019 | |||
BC026896.1 | - | 3 | 360 | |||
BC141210.1 | - | 18 | 1970 | 9055822 |