Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199027.2 | NP_950192.2 | 14 | 2174 | ||
GenBank mRNA | AK157577.1 | - | 14 | 2005 | ||
BC062921.1 | - | 13 | 2097 | |||
BC151013.1 | - | 13 | 1914 | 9087926 | ||
BC157954.1 | - | 13 | 1914 | 9088127 | ||
BC172135.1 | - | 12 | 1993 | |||
CN721033.1 | - | 3 | 200 |