Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183319.2 | NP_899142.2 | 2 | 432 | ||
GenBank mRNA | AY534249.1 | - | 2 | 432 | ||
BC150896.1 | - | 2 | 468 | 9087807 | ||
BC150897.1 | - | 2 | 468 | 9087808 |