Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 6, 7 | 956 | ||
GenBank mRNA | AK085308.1 | - | 6, 7 | 946 | ||
AK165706.1 | - | 6, 7 | 908 | |||
BC043305.1 | - | 6, 7 | 885 | 5040944 |