Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011584.4 | NP_035714.3 | 7 | 1720 | ||
GenBank mRNA | AK033326.1 | - | 2 | 212 | ||
AK079075.1 | - | 7 | 1607 | |||
BC008556.1 | - | 12 | ||||
BC040082.1 | - | 12 | ||||
BC096461.1 | - | 7 | 1560 | 5361394 | ||
U09504.1 | - | 6 | 1338 | |||
U12142.1 | - | 7 | 1686 |