Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291212.1 | NP_001278141.1 | 34 | 5217 | ||
NM_001291213.1 | NP_001278142.1 | 34 | 5325 | |||
NM_001291214.1 | NP_001278143.1 | 32 | 4172 | |||
NM_001291215.1 | NP_001278144.1 | 35 | 4238 | |||
NM_011586.3 | NP_035716.1 | 33 | 5181 | |||
GenBank mRNA | AB026497.1 | - | 33 | 5151 | ||
AK147584.1 | - | 35 | 4187 | |||
AK171342.1 | - | 32 | 4169 | |||
BC023378.1 | - | 9 | 1046 | |||
BC046638.1 | - | 16 | 2009 | |||
BC057920.1 | - | 4 | 392 | |||
BC138366.1 | - | 34 | 5159 | 8861386 | ||
BC145257.1 | - | 34 | 5267 |