Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 10 | 1794 | ||
GenBank mRNA | AF422244.1 | - | 10 | 1731 | ||
AF422245.1 | - | 10 | 1731 | |||
AK085904.1 | - | 10 | 1690 | |||
AK137442.1 | - | 10 | 1709 | |||
BC141023.1 | - | 10 | 1707 | 9055635 |