Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033439.3 | NP_001028611.2 | 12 | 1591 | ||
NM_001252132.1 | NP_001239061.1 | 12 | 1591 | |||
GenBank mRNA | AK076431.1 | - | 3 | 190 | ||
AK129264.1 | - | 12 | 1580 | |||
AK136026.1 | - | 12 | 1197 | |||
AK143909.1 | - | 12 | 1590 | |||
AK160902.1 | - | 10 | 967 | |||
AK162105.1 | - | 2 | 98 |