Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201529.2 | NP_963287.2 | 25, 26 | 4542 | ||
GenBank mRNA | AK047836.1 | - | 1, 2 | 1471 | ||
AK129231.1 | - | 22, 23 | 4327 | |||
AK137694.1 | - | 16, 17 | 2781 | |||
BC082553.1 | - | 23, 24 | 4557 | |||
BC145011.1 | - | 18, 19 | 3026 | 9053559 | ||
EF071857.1 | - | 25, 26 | 4513 |