Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 5 | 503 | ||
NM_001289485.1 | NP_001276414.1 | 6 | 588 | |||
NM_001289486.1 | NP_001276415.1 | 5 | 503 | |||
GenBank mRNA | AF338468.1 | - | 3 | 267 | ||
AF338469.1 | - | 6 | 582 | |||
BB618060.1 | - | 5 | 504 | |||
BC050244.1 | - | 3 | 287 | |||
BC150885.1 | - | 5 | 465 | 9087796 | ||
BC172032.1 | - | 5 | 465 | 9054318 |