Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 8, 9 | 942 | ||
NM_001289485.1 | NP_001276414.1 | 9, 10 | 1027 | |||
NM_001289486.1 | NP_001276415.1 | 8, 9 | 939 | |||
GenBank mRNA | AF338468.1 | - | 6, 7 | 706 | ||
AF338469.1 | - | 9, 10 | 1021 | |||
AK053319.1 | - | 2, 3 | 276 | |||
BC050244.1 | - | 6, 7 | 727 | |||
BC150885.1 | - | 8, 9 | 904 | 9087796 | ||
BC172032.1 | - | 8, 9 | 901 | 9054318 |