Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033449.1 | NP_001028621.1 | 5, 6 | 393 | ||
NM_001037812.2 | NP_001032901.1 | 5, 6 | 352 | |||
GenBank mRNA | AK133112.1 | - | 6, 7 | 377 | ||
AK161697.1 | - | 5, 6 | 395 | |||
AK167733.1 | - | 5, 6 | 354 | |||
AK170118.1 | - | 5, 6 | 333 | |||
BC147243.1 | - | 5, 6 | 372 | 9056304 | ||
BC147244.1 | - | 5, 6 | 372 | 9056305 | ||
BF722386.1 | - | 1, 2 | 39 |