Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081201.1 | NP_001074670.1 | 14, 15 | 1683 | ||
GenBank mRNA | AK144417.1 | - | 14, 15 | 1635 | ||
AK172656.1 | - | 13, 14 | 1559 | |||
BC147500.1 | - | 14, 15 | 1639 | 9056363 | ||
BC157932.1 | - | 14, 15 | 1639 | 9088105 | ||
BC157988.1 | - | 14, 15 | 1639 | 9088143 |