Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033473.2 | NP_001028645.1 | 3 | 316 | ||
GenBank mRNA | AK158063.1 | - | 3 | 317 | ||
BC147356.1 | - | 3 | 304 | 9056405 | ||
BC147357.1 | - | 3 | 304 | 9056398 | ||
BC172043.1 | - | 2 | 196 | 9054329 |