Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027740.6 | NP_082016.1 | 5 | 569 | ||
GenBank mRNA | AK016991.1 | - | 4 | 493 | ||
AK030180.1 | - | 4 | 463 | |||
AK045302.1 | - | 5 | 548 | |||
AK050233.1 | - | 5 | 638 | |||
AK090075.1 | - | 5 | 572 | |||
BC050888.1 | - | 4 | 567 | |||
BC099926.1 | - | 5 | 656 | |||
BC144819.1 | - | 5 | 658 | |||
BC145801.1 | - | 5 | 658 | 40131104 |