Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028260.2 | NP_082536.1 | 7 | 692 | ||
GenBank mRNA | AK005356.1 | - | 7 | 693 | ||
AK010075.1 | - | 7 | 650 | |||
AK011278.1 | - | 7 | 693 | |||
AK011382.1 | - | 7 | 693 | |||
AK011420.1 | - | 7 | 654 | |||
AK011466.1 | - | 7 | 665 | |||
AK011618.1 | - | 7 | 654 | |||
AK011872.1 | - | 7 | 656 | |||
AK012181.1 | - | 645 | ||||
AK015978.1 | - | 7 | 663 | |||
AK078300.1 | - | 7 | 679 | |||
AK135670.1 | - | 7 | 652 | |||
AK168202.1 | - | 7 | 641 | |||
BC008259.1 | - | 7 | 651 | 2649406 | ||
BC081433.1 | - | 6 | 535 | 6814816 |