Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007472.2 | NP_001007473.1 | 3 | 714 | ||
GenBank mRNA | AJ862873.1 | - | 3 | 671 | ||
AJ862874.1 | - | 3 | 671 | |||
AJ862875.1 | - | 3 | 671 | |||
AK135886.1 | - | 3 | 714 | |||
BC139154.1 | - | 3 | 658 | 8862176 | ||
BC139155.1 | - | 3 | 658 | 8862177 |