Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165968.1 | NP_001159440.1 | 2 | 152 | ||
NM_207263.2 | NP_997146.2 | 2 | 152 | |||
GenBank mRNA | AY518699.1 | - | 2 | 106 | ||
BC120840.1 | - | 2 | 147 | 40129763 | ||
BC144902.1 | - | 2 | 152 | 9053450 | ||
BC144927.1 | - | 2 | 152 |