Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270537.1 | NP_001257466.1 | 8 | 657 | ||
GenBank mRNA | BC114419.1 | - | 1 | 507 | ||
BC127021.1 | - | 1 | 508 | |||
BC141885.1 | - | 1 | 508 | |||
CN691011.1 | - | 4 | 308 |