Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027976.2 | NP_082252.1 | 10 | 1640 | ||
GenBank mRNA | AK034942.1 | - | 1640 | |||
AK145894.1 | - | 10 | 1301 | |||
AK150542.1 | - | 10 | 1306 | |||
AK150663.1 | - | 10 | 1306 | |||
AK151378.1 | - | 10 | 1095 | |||
AK161024.1 | - | 10 | 993 | |||
AK206020.1 | - | 3 | 107 | |||
AK219510.1 | - | 3 | 107 | |||
BC031544.1 | - | 10 | 1074 | 3987201 |