Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013806.1 | NP_001013828.1 | 12 | 1482 | ||
GenBank mRNA | AK135906.1 | - | 13 | 1218 | ||
AK157542.1 | - | 1664 | ||||
BC065088.1 | - | 11 | 3676 | |||
BC138991.1 | - | 12 | 1490 | 8862013 | ||
BC138992.1 | - | 13 | 1469 | 8862014 | ||
CF736962.1 | - | 5 | 478 |