Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 15, 16 | 1774 | ||
NM_028027.3 | NP_082303.2 | 15, 16 | 1836 | |||
GenBank mRNA | AF487515.1 | - | 15, 16 | 1802 | ||
AK010452.1 | - | 15, 16 | 1802 | |||
AK158684.1 | - | 15, 16 | 1774 | |||
BC023367.1 | - | 6, 7 | 813 | |||
BC041113.1 | - | 13, 14 | 2150 | |||
BC054833.1 | - | 6, 7 | 1297 | 6439728 |