Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 14, 15 | 1554 | ||
NM_028027.3 | NP_082303.2 | 14, 15 | 1616 | |||
GenBank mRNA | AF487515.1 | - | 14, 15 | 1582 | ||
AK010452.1 | - | 14, 15 | 1582 | |||
AK158684.1 | - | 14, 15 | 1554 | |||
BC023367.1 | - | 5, 6 | 593 | |||
BC041113.1 | - | 12, 13 | 1930 |