Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020622.2 | NP_065647.1 | 4 | 427 | ||
GenBank mRNA | AF360358.1 | - | 4 | 428 | ||
AF494380.1 | - | 4 | 332 | |||
AK018571.1 | - | 4 | 397 | |||
BC048949.1 | - | 4 | 374 | 6474753 |