Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159706.1 | NP_001153178.1 | 17 | 2035 | ||
NM_016770.3 | NP_058050.3 | 18 | 2134 | |||
GenBank mRNA | AF026380.1 | - | 18 | 2116 | ||
AK002920.1 | - | 18 | 2101 | |||
AK090289.1 | - | 2137 | ||||
BC119604.1 | - | 17 | 1979 | 40060385 | ||
BC119605.1 | - | 18 | 2078 | 40060386 |