Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291012.1 | NP_001277941.1 | 19 | 1810 | ||
NM_001291013.1 | NP_001277942.1 | 19 | 1807 | |||
NM_016902.4 | NP_058598.1 | 19 | 1813 | |||
GenBank mRNA | AF127180.1 | - | 19 | 1764 | ||
AF176210.1 | - | 19 | 1776 | |||
AJ243223.1 | - | 19 | 1761 | |||
AK009252.1 | - | 2 | 149 | |||
AK159627.1 | - | 19 | 1787 | |||
BC118953.2 | - | 19 | 1758 | 40093235 | ||
EF174553.3 | - | 19 | 1782 |