Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291012.1 | NP_001277941.1 | 20 | 2078 | ||
NM_001291013.1 | NP_001277942.1 | 20 | 2075 | |||
NM_016902.4 | NP_058598.1 | 20 | 2081 | |||
GenBank mRNA | AA107655.1 | - | 1 | 132 | ||
AF127180.1 | - | 20 | 2032 | |||
AF176210.1 | - | 20 | 2044 | |||
AJ243223.1 | - | 20 | 2029 | |||
AK009252.1 | - | 3 | 417 | |||
AK159627.1 | - | 20 | 2055 | |||
BC118953.2 | - | 20 | 2026 | 40093235 | ||
EF174553.3 | - | 20 | 2050 | |||
EF174554.2 | - | 5 | 466 | |||
FJ767895.1 | - | 5 | 451 | |||
FJ767896.1 | - | 6 | 492 | |||
FJ767897.1 | - | 5 | 456 | |||
FJ767898.1 | - | 5 | 456 |