Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 20 | 2183 | ||
GenBank mRNA | AB037111.1 | - | 21 | 2468 | ||
AB037181.1 | - | 20 | 2183 | |||
AK080951.1 | - | 20 | 2360 | |||
BC022604.1 | - | 11 | 965 | |||
BC137898.1 | - | 20 | 2166 | 8860917 | ||
BC145016.1 | - | 20 | 2166 | 9053564 |