Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290455.1 | NP_001277384.1 | 14 | 1130 | ||
NM_207670.2 | NP_997553.1 | 13 | 1037 | |||
GenBank mRNA | AK037997.1 | - | 7 | 1492 | ||
AK156165.1 | - | 13 | 1038 | |||
AK158098.1 | - | 10 | 835 | |||
AK172590.1 | - | 11 | 903 | |||
AK172623.1 | - | 13 | 1038 | |||
AK172699.1 | - | 13 | 1038 | |||
AK173119.1 | - | 1 | 53 | |||
AW342118.1 | - | 13 | 1012 | |||
BC055931.1 | - | 2 | 144 | |||
BC138523.1 | - | 14 | 1126 | 8861543 | ||
BC145310.1 | - | 14 | 1126 | 9053864 |