Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019635.2 | NP_062609.2 | 4 | 577 | ||
GenBank mRNA | AF271361.1 | - | 4 | 323 | ||
AY058922.1 | - | 4 | 463 | |||
BC037440.1 | - | 2 | 333 | 3585312 | ||
BC049123.1 | - | 4 | 451 | 6813956 | ||
U28726.1 | - | 4 | 456 |