Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019662.2 | NP_062636.2 | 3 | 685 | ||
GenBank mRNA | AF084466.1 | - | 2 | 428 | ||
AK077271.1 | - | 3 | 683 | |||
AK087473.1 | - | 3 | 658 | |||
AK159550.1 | - | 2 | 468 | |||
BC057138.1 | - | 3 | 664 | 5149047 |