Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019649.2 | NP_062623.2 | 9 | 1095 | ||
GenBank mRNA | AK148080.1 | - | 9 | 1077 | ||
AK152410.1 | - | 9 | 1085 | |||
AK152809.1 | - | 9 | 1085 | |||
AK165742.1 | - | 8 | 1789 | |||
AK171901.1 | - | 9 | 1086 | |||
AK191893.1 | - | 2 | 158 | |||
AK205055.1 | - | 2 | 135 | |||
BC022172.1 | - | 9 | 1086 | 4193279 | ||
D67067.1 | - | 9 | 1082 |