Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285891.1 | NP_001272820.1 | 7 | 903 | ||
NM_001285892.1 | NP_001272821.1 | 7 | 903 | |||
NM_019748.3 | NP_062722.1 | 7 | 903 | |||
GenBank mRNA | AB024303.1 | - | 7 | 791 | ||
AK010313.1 | - | 7 | 780 | |||
AK011772.1 | - | 7 | 814 | |||
AK011783.1 | - | 4 | 250 | |||
AK087556.1 | - | 7 | 794 | |||
AK090012.1 | - | 7 | 790 | |||
AK154139.1 | - | 7 | 816 | |||
AK159672.1 | - | 7 | 805 | |||
AK162789.1 | - | 7 | 791 | |||
BC068164.1 | - | 7 | 792 | 30359189 |