Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019776.2 | NP_062750.2 | 15 | 1809 | ||
GenBank mRNA | AB021491.1 | - | 15 | 1766 | ||
AK034914.1 | - | 14 | 1678 | |||
AK088028.1 | - | 15 | 1768 | |||
AK133837.1 | - | 15 | 1815 | |||
AK159837.1 | - | 15 | 1774 | |||
AK161591.1 | - | 15 | 1763 | |||
AK162432.1 | - | 15 | 1750 | |||
AK167576.1 | - | 15 | 1799 | |||
BC007126.1 | - | 15 | 1756 | 3592548 |