Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190320.1 | NP_001177249.1 | 4 | 480 | ||
NM_001190321.1 | NP_001177250.1 | 4 | 468 | |||
NM_020001.2 | NP_064385.1 | 5 | 570 | |||
GenBank mRNA | AF240357.1 | - | 5 | 539 | ||
AF240358.1 | - | 4 | 437 | |||
AF240359.1 | - | 5 | 539 | |||
AK089166.1 | - | 5 | 557 | |||
AK131697.1 | - | 5 | 557 | |||
AK154673.1 | - | 4 | 467 | |||
AK171046.1 | - | 6 | 740 | |||
BC023008.1 | - | 5 | 539 | 5376247 |