Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019993.3 | NP_064377.2 | 7 | 1249 | ||
GenBank mRNA | AB296386.1 | - | 7 | 1194 | ||
AF170919.1 | - | 7 | 994 | |||
AK132336.1 | - | 7 | 1034 | |||
AK149589.1 | - | 7 | 1035 | |||
AK150093.1 | - | 7 | 992 | |||
AK154914.1 | - | 7 | 1233 | |||
AK162354.1 | - | 7 | 1033 | |||
AK165730.1 | - | 7 | 1249 | |||
AK168261.1 | - | 7 | 1227 | |||
AK168309.1 | - | 7 | 1227 | |||
AK168878.1 | - | 7 | 1198 | |||
AK179746.1 | - | 2 | 200 | |||
AK184027.1 | - | 2 | 200 | |||
AK185844.1 | - | 1 | 40 | |||
AK188805.1 | - | 2 | 200 | |||
AK199575.1 | - | 2 | 200 | |||
AK203750.1 | - | 2 | 200 | |||
BC003297.1 | - | 7 | 1026 | 3583976 |