Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020560.2 | NP_065585.1 | 3 | 582 | ||
GenBank mRNA | AK005110.1 | - | 3 | 541 | ||
AK011825.1 | - | 3 | 538 | |||
AK019202.1 | - | 3 | 562 | |||
AK078073.1 | - | 3 | 525 | |||
AK161913.1 | - | 3 | 525 | |||
BC006764.1 | - | 3 | 477 | |||
BC027430.1 | - | 3 | 486 | 4187438 | ||
Z46966.1 | - | 3 | 501 |