Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB162929.1 | - | 3 | 326 | ||
AF190699.1 | - | 3 | 398 | |||
AF190991.1 | - | 3 | 398 | |||
AK015486.1 | - | 2 | 49 | |||
AK018148.1 | - | 3 | 390 | |||
AK031550.1 | - | 3 | 426 | |||
AK042758.1 | - | 3 | 179 | |||
AK043998.1 | - | 3 | 446 | |||
AK048413.1 | - | 3 | 397 | |||
AK049903.1 | - | 2 | 132 | |||
AK081981.1 | - | 3 | 399 | |||
AK083126.1 | - | 2 | 523 | |||
BC046310.1 | - | 3 | 384 | 6307420 | ||
BC065694.1 | - | 3 | 390 | 6813365 |